Thermo Scientific Chemicals

Selenium(IV) oxide, 99.8%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 200000100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Selenium(IV) oxide, 99.8%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 200000100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 200000100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7446-08-4
IUPAC Name
(oxo-λ⁴-selanylidene)oxidane
Molecular Formula
O2Se
InChI Key
JPJALAQPGMAKDF-UHFFFAOYSA-N
SMILES
O=[Se]=O
Appearance (Form)
Powder
Appearance (Color)
White to light beige
Total trace metal impurities
=<0.2 %
Iron (Fe)
=<50 ppm
Copper (Cu)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text