Thermo Scientific Chemicals

4-Pyrrolidinopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 200010050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Pyrrolidinopyridine, 98%, Thermo Scientific Chemicals

Catalog number: 200010050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 200010050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2456-81-7
IUPAC Name
4-(pyrrolidin-1-yl)pyridin-1-ium
Molecular Formula
C9H13N2
InChI Key
RGUKYNXWOWSRET-UHFFFAOYSA-O
SMILES
C1CCN(C1)C1=CC=[NH+]C=C1
Appearance (Color)
White to yellow to light brown
Appearance (Form)
Crystals or crystalline mass and/or chunks
Melting point
54°C to 58°C
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text