Pyronin Y, pure, high purity biological stain
Pyronin Y, pure, high purity biological stain
Thermo Scientific Chemicals

Pyronin Y, pure, high purity biological stain

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 200100010
Price (USD)/ Each
37.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
44.70 
Save 7.05 (16%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
37.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
44.70 
Save 7.05 (16%)
Add to cart
Pyronin Y, pure, high purity biological stain
Catalog number200100010
Price (USD)/ Each
37.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
44.70 
Save 7.05 (16%)
-
Add to cart
Chemical Identifiers
CAS92-32-0
IUPAC Name3,6-bis(dimethylamino)-10λ⁴-xanthen-10-ylium chloride
Molecular FormulaC17H19ClN2O
InChI KeyINCIMLINXXICKS-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC2=[O+]C3=CC(=CC=C3C=C2C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Absorption Ratio P-15/P+151.23 to 1.7
Dye content>=50 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only