Thermo Scientific Chemicals

Dimethylcyanamide, 97%, Thermo Scientific Chemicals

Catalog number: 200211000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Dimethylcyanamide, 97%, Thermo Scientific Chemicals

Catalog number: 200211000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 200211000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1467-79-4
IUPAC Name
cyanodimethylamine
Molecular Formula
C3H6N2
InChI Key
OAGOUCJGXNLJNL-UHFFFAOYSA-N
SMILES
CN(C)C#N
Water
=<0.1 % (K.F.)
Refractive index
1.4090 to 1.4110 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text