Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals
Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 200330250
Price (USD)/ Each
108.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
128.00 
Save 19.35 (15%)
-
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
108.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
128.00 
Save 19.35 (15%)
Add to cart
Vinyltrimethylsilane, 97%, Thermo Scientific Chemicals
Catalog number200330250
Price (USD)/ Each
108.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
128.00 
Save 19.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameethenyltrimethylsilane
Molecular FormulaC5H12Si
InChI KeyGCSJLQSCSDMKTP-UHFFFAOYSA-N
SMILESC[Si](C)(C)C=C
Molecular Weight (g/mol)100.24
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.3895 to 1.3915 (20°C, 589 nm)
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only