Thermo Scientific Chemicals

4-Iodopyrazole, 98%, Thermo Scientific Chemicals

Catalog number: 200370250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Iodopyrazole, 98%, Thermo Scientific Chemicals

Catalog number: 200370250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 200370250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3469-69-0
IUPAC Name
4-iodo-1H-pyrazole
Molecular Formula
C3H3IN2
InChI Key
LLNQWPTUJJYTTE-UHFFFAOYSA-N
SMILES
IC1=CNN=C1
Appearance (Form)
Crystalline powder
HPLC
>=97.5 %
Infrared spectrum
Conforms
Melting point
107°C to 112°C
Appearance (Color)
White to off-white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text