Thermo Scientific Chemicals

2,2,6,6-Tetramethylpiperidinooxy, 98%, Thermo Scientific Chemicals

Catalog number: 200380250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2,2,6,6-Tetramethylpiperidinooxy, 98%, Thermo Scientific Chemicals

Catalog number: 200380250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 200380250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2564-83-2
IUPAC Name
(2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Molecular Formula
C9H18NO
InChI Key
QYTDEUPAUMOIOP-UHFFFAOYSA-N
SMILES
CC1(C)CCCC(C)(C)N1[O]
Appearance (Form)
Crystalline powder or crystals or solid and/or
Infrared spectrum
Conforms
Appearance (Color)
Orange to red
Appearance (Form)
chunks
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text