Benzylamine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Benzylamine hydrochloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
View all
Plastic bottle
Catalog number 200541000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS3287-99-8
IUPAC Namephenylmethanaminium chloride
Molecular FormulaC7H10ClN
InChI KeyXKXHCNPAFAXVRZ-UHFFFAOYSA-N
SMILES[Cl-].[NH3+]CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point259°C to 264°C
Titration with HClO4>=98.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only