Thermo Scientific Chemicals

Sodium selenite, 44-46% Se, anhydrous, Thermo Scientific Chemicals

Catalog number: 200730250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium selenite, 44-46% Se, anhydrous, Thermo Scientific Chemicals

Catalog number: 200730250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 200730250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10102-18-8
IUPAC Name
disodium selenite
Molecular Formula
Na2O3Se
InChI Key
BVTBRVFYZUCAKH-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-][Se]([O-])=O
Lead (Pb)
=<500 ppm
Appearance (Form)
Powder
Loss on drying
=<0.5 %
Iron (Fe)
=<300 ppm
Assay
44 to 46 % (Se)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text