Sodium selenite, 44-46% Se, anhydrous, Thermo Scientific Chemicals
Sodium selenite, 44-46% Se, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium selenite, 44-46% Se, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 200735000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10102-18-8
IUPAC Namedisodium selenite
Molecular FormulaNa2O3Se
InChI KeyBVTBRVFYZUCAKH-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-][Se]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay44 to 46 % (Se)
Lead (Pb)=<500 ppm
Appearance (Form)Powder
Loss on drying=<0.5 %
Iron (Fe)=<300 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only