Thermo Scientific Chemicals

Chrysoidin, pure

Catalog number: 200740500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Chrysoidin, pure

Catalog number: 200740500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 200740500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
532-82-1
IUPAC Name
hydrogen 4-(2-phenyldiazen-1-yl)benzene-1,3-diamine chloride
Molecular Formula
C12H13ClN4
InChI Key
MCTQNEBFZMBRSQ-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC1=CC=C(N=NC2=CC=CC=C2)C(N)=C1
Appearance (Color)
Bordeaux to purple
Infrared spectrum
Conforms
Visual transition interval
from pH 4.0 (orange) to pH 7.0 (yellow)
UV spectrum
Complies
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text