Amino Acid Standard H
Amino Acid Standard H
Thermo Scientific™

Amino Acid Standard H

Thermo Scientific Pierce Amino Acid Standard H is a quantitative mixture of 18 amino acids, supplied at 2.5 μMol/mL eachRead more
Have Questions?
Catalog NumberQuantity
2008810 x 1 mL
Catalog number 20088
Price (KRW)
241,000
Each
-
Add to cart
Quantity:
10 x 1 mL
Request bulk or custom format
Price (KRW)
241,000
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce Amino Acid Standard H is a quantitative mixture of 18 amino acids, supplied at 2.5 μMol/mL each in 0.1N HCl, for use as a high-purity calibration standard for HPLC analysis of protein hydrolysates.

Features of Amino Acid Standard H:

Complete mixture—contains eighteen (18) amino acids for assessment of amino acid composition of peptide hydrolysates
Quantitative—contains exact amounts of each amino acid (2.5±0.1 μMol/mL, except cystine at 1.25±0.1 μMol/mL), enabling its use as a quantitation standard
Stable—dissolved in 0.1N hydrochloric acid and stabilized as 1 mL aliquots in sealed glass ampules
Standard H—designed for compositional amino acid analysis by HPLC methods of protein or peptide hydrolysates

This quantitative solution of eighteen purified amino acids in 0.1N HCl is an ideal reference standard for calibrating amino acid analyzers and assessing the composition of hydrolyzed peptides and proteins (hydrolysates). To permit standardization of microbiological and other assays, the mixture uses only the L-form of each amino acid. The molar concentration of each amino acid is verified by conventional amino acid analysis methods, making the mixture suitable as both a qualitative and quantitative standard. The solution is provided as stable 1 mL aliquots in sealed glass ampules.

Specifications:
Product Details:
When kept frozen, an unopened vial has an indefinite storage life. Once the seal is broken, the reagent has a maximum storage life of six months (store frozen between uses). To use the product, thaw a vial and shake well to ensure a homogeneous solution. Then dilute appropriately with suitable buffers to a concentration compatible with the full-scale sensitivity of your amino acid analyzer.

For added safety when opening glass ampules, consider using our Ampule Breakers, which are disposable safety devices that protect the fingers when breaking open a glass ampule.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 x 1 mL
FormLiquid
Product TypeAmino Acid Standard H
Unit SizeEach
Contents & Storage
Store between -20°C and 8°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link