Thermo Scientific Chemicals

Pyridinium dichromate, 98%, Thermo Scientific Chemicals

Catalog number: 200880010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Pyridinium dichromate, 98%, Thermo Scientific Chemicals

Catalog number: 200880010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 200880010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20039-37-6
IUPAC Name
[(hydroxydioxochromio)oxy]chromiumoylol; bis(pyridine)
Molecular Formula
C10H12Cr2N2O7
InChI Key
RCBVKBFIWMOMHF-UHFFFAOYSA-L
SMILES
C1=CC=NC=C1.C1=CC=NC=C1.O[Cr](=O)(=O)O[Cr](O)(=O)=O
Titration Iodometric
>=97.5 %
Appearance (Color)
Orange to brown
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text