1x1 image pixel for data collection
Pyridinium dichromate, 98%
Pyridinium dichromate, 98%
Pyridinium dichromate, 98%
Pyridinium dichromate, 98%
Thermo Scientific Chemicals

Pyridinium dichromate, 98%

CAS: 20039-37-6 | C10H12Cr2N2O7 | 376.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 200882500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS20039-37-6
IUPAC Name[(hydroxydioxochromio)oxy]chromiumoylol; bis(pyridine)
Molecular FormulaC10H12Cr2N2O7
InChI KeyRCBVKBFIWMOMHF-UHFFFAOYSA-L
SMILESC1=CC=NC=C1.C1=CC=NC=C1.O[Cr](=O)(=O)O[Cr](O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodometric>=97.5 %
Appearance (Color)Orange to brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection