1x1 image pixel for data collection
Phosphorus pentoxide, 98%, extra pure
Phosphorus pentoxide, 98%, extra pure
Phosphorus pentoxide, 98%, extra pure
Phosphorus pentoxide, 98%, extra pure
Thermo Scientific Chemicals

Phosphorus pentoxide, 98%, extra pure

CAS: 1314-56-3 | O5P2 | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 200890025
Price (USD)
224.65
Online Exclusive
249.00
Save 24.35 (10%)
Each
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
224.65
Online Exclusive
249.00
Save 24.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1314-56-3
IUPAC Nametricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
Molecular FormulaO5P2
InChI KeyDLYUQMMRRRQYAE-UHFFFAOYSA-N
SMILESO=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
View more
SpecificationsSpecification SheetSpecification Sheet
Reducing substance (P2O3)=<0.1 %
Appearance (Color)White
Titration with NaOH>=97.5 %
Appearance (Form)Hygroscopic crystalline powder
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0471863Certificate of AnalysisMay 17, 2025200890010, 200890025, 200892500, 200890100
A0468170Certificate of AnalysisJan 31, 2025200890010, 200890025, 200892500, 200890100
A0444728Certificate of AnalysisJan 31, 2025200890010, 200890025, 200892500, 200890100
A0466878Certificate of AnalysisOct 17, 2024200890010, 200890025, 200892500, 200890100
A0461486Certificate of AnalysisMar 31, 2024200890010, 200890025, 200892500, 200890100
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection