Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
250 g
1 kg
2.5 kg
Packaging:
View all
Plastic drum
Catalog number 200890100
Price (USD)/ Each
618.65
Online exclusive
687.00 
Save 68.35 (10%)
-
Add to cart
Quantity:
10 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)/ Each
618.65
Online exclusive
687.00 
Save 68.35 (10%)
Add to cart
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Catalog number200890100
Price (USD)/ Each
618.65
Online exclusive
687.00 
Save 68.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-56-3
IUPAC Nametricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
Molecular FormulaO5P2
InChI KeyDLYUQMMRRRQYAE-UHFFFAOYSA-N
SMILESO=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
View more
SpecificationsSpecification SheetSpecification Sheet
Reducing substance (P2O3)=<0.1 %
Appearance (Color)White
Titration with NaOH>=97.5 %
Appearance (Form)Hygroscopic crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only