Thermo Scientific Chemicals

Diisobutylaluminium hydride, 1.2M (20 wt%) solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 201084000
400 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Diisobutylaluminium hydride, 1.2M (20 wt%) solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 201084000
400 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 201084000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1191-15-7, 108-88-3
Appearance (Form)
Clear solution
Strength
19.5 to 20.5 % (DIBAL-H, on Al basis)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid, Reduction reagent
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text