Thermo Scientific Chemicals

Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 201100050
5 g, Plastic bottle, Each
Thermo Scientific Chemicals

Gallium(III) oxide, 99.99+%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 201100050
5 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 201100050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12024-21-4
IUPAC Name
digallium(3+) trioxidandiide
Molecular Formula
Ga2O3
InChI Key
AJNVQOSZGJRYEI-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Ga+3].[Ga+3]
Appearance (Color)
White
Total trace metal impurities
=<0.010 %
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text