Potassium cyanate, 97%, pure
Potassium cyanate, 97%, pure
Potassium cyanate, 97%, pure
Potassium cyanate, 97%, pure
Thermo Scientific Chemicals

Potassium cyanate, 97%, pure

CAS: 590-28-3 | CKNO | 81.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
100 g
500 g
Packaging:
Plastic bottle
Catalog number 201111000
Price (USD)
36.65
Special offer
Online exclusive
Ends: 26-Sep-2025
44.50
Save 7.85 (18%)
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
36.65
Special offer
Online exclusive
Ends: 26-Sep-2025
44.50
Save 7.85 (18%)
Each
Add to cart
Chemical Identifiers
CAS590-28-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration Argentometric>=96.0 %
Appearance (Form)Crystalline powder and/or chunks
Potassium carbonate=<3 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link