Thermo Scientific Chemicals

Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 201150100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Phosphorous acid, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 201150100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 201150100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13598-36-2
IUPAC Name
dihydroxy(oxo)-λ⁵-phosphanylium
Molecular Formula
H2O3P
InChI Key
XNQULTQRGBXLIA-UHFFFAOYSA-O
SMILES
O[P+](O)=O
Appearance (Color)
White
Appearance (Form)
Crystals
Titration with NaOH
97.5 to 102.5 %
Heavy metals (as Pb)
=<20 ppm
Chloride (Cl)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text