Calcium, 99%, granular, Thermo Scientific Chemicals
Calcium, 99%, granular, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium, 99%, granular, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Plastic Bottle
Catalog number 201180050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-70-2
IUPAC Namecalcium
Molecular FormulaCa
InChI KeyOYPRJOBELJOOCE-UHFFFAOYSA-N
SMILES[Ca]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Metallic pieces
Assay>=98.5 %
Appearance (Color)Grey
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only