Thermo Scientific Chemicals

Antimony(V)-chloride, 99%, anhydrous, Thermo Scientific Chemicals

Catalog number: 201210500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Antimony(V)-chloride, 99%, anhydrous, Thermo Scientific Chemicals

Catalog number: 201210500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 201210500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7647-18-9
IUPAC Name
antimony pentachloride
Molecular Formula
Cl5Sb
InChI Key
VMPVEPPRYRXYNP-UHFFFAOYSA-I
SMILES
Cl[Sb](Cl)(Cl)(Cl)Cl
Appearance (Color)
Colorless to yellow to orange
Appearance (Form)
Liquid
Appearance
May develop some turbidity or precipitate
Titration Iodometric
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text