Thermo Scientific Chemicals

Rubidium chloride, 99+%, Thermo Scientific Chemicals

Catalog number: 201271000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Rubidium chloride, 99+%, Thermo Scientific Chemicals

Catalog number: 201271000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 201271000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7791-11-9
IUPAC Name
rubidium(1+) chloride
Molecular Formula
ClRb
InChI Key
FGDZQCVHDSGLHJ-UHFFFAOYSA-M
SMILES
[Cl-].[Rb+]
Appearance (Color)
White
Appearance (Form)
Powder or crystals and/or chunks
Total trace metal impurities
=<1.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text