Thermo Scientific Chemicals

1-Methyladenine, 98+%

Catalog number: 201311000
100 mg, Glass bottle, Each
Thermo Scientific Chemicals

1-Methyladenine, 98+%

Catalog number: 201311000
100 mg, Glass bottle, Each
Quantity
Packaging
Catalog number: 201311000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5142-22-3
IUPAC Name
1-methyl-1H-purin-6-amine
Molecular Formula
C6H7N5
InChI Key
HPZMWTNATZPBIH-UHFFFAOYSA-N
SMILES
CN1C=NC2=NC=NC2=C1N
Appearance (Color)
Light yellow to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
HPLC
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text