Thermo Scientific Chemicals

Acenaphthene, 99%, Thermo Scientific Chemicals

Catalog number: 201341000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Acenaphthene, 99%, Thermo Scientific Chemicals

Catalog number: 201341000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 201341000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
83-32-9
IUPAC Name
1,2-dihydroacenaphthylene
Molecular Formula
C12H10
InChI Key
CWRYPZZKDGJXCA-UHFFFAOYSA-N
SMILES
C1CC2=C3C1=CC=CC3=CC=C2
Appearance (Color)
White to yellow to brown
Appearance (Form)
Crystalline powder or crystals and/or chunks
Infrared spectrum
Conforms
Melting point
91°C to 95°C
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text