Thermo Scientific Chemicals

Copper(II) perchlorate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 201361000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Copper(II) perchlorate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 201361000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 201361000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10294-46-9
IUPAC Name
copper(2+) hexahydrate diperchlorate
Molecular Formula
Cl2CuH12O14
InChI Key
NHELIHXBJRANPL-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cu++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Appearance (Form)
Adhering crystals
Chloride (Cl)
=<100 ppm
Sulfate (SO4)
=<50 ppm
Titration Iodometric
>=97.5 %
Insoluble matter
=<0.01 % (in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text