Thermo Scientific Chemicals

Ammonium iron(II) sulfate hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 201370050
5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium iron(II) sulfate hexahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 201370050
5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 201370050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7783-85-9
IUPAC Name
λ²-iron(2+) diammonium hexahydrate disulfate
Molecular Formula
FeH20N2O14S2
InChI Key
MQLVWQSVRZVNIP-UHFFFAOYSA-L
SMILES
[NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Appearance (Color)
Light green
Appearance (Form)
Crystalline powder or crystals
Copper (Cu)
=<20 ppm
Zinc (Zn)
=<30 ppm
Ferric iron (Fe 3+)
=<200 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text