Iron(II) sulfate heptahydrate, 99.5%, for analysis
Iron(II) sulfate heptahydrate, 99.5%, for analysis
Iron(II) sulfate heptahydrate, 99.5%, for analysis
Iron(II) sulfate heptahydrate, 99.5%, for analysis
Thermo Scientific Chemicals

Iron(II) sulfate heptahydrate, 99.5%, for analysis

CAS: 7782-63-0 | FeH14O11S | 278.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 201392500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7782-63-0
IUPAC Nameλ²-iron(2+) heptahydrate sulfate
Molecular FormulaFeH14O11S
InChI KeySURQXAFEQWPFPV-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Copper (Cu)=<10 ppm
Appearance (Color)Light green to blue
Subst not pptd by NH3=<0.05 % (as SO4)
Total nitrogen compounds=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link