Chemical Identifiers
CAS16774-21-3
IUPAC Nameλ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
Molecular FormulaCeH8N8O18
InChI KeyWIBGOERAEYJBOT-UHFFFAOYSA-N
SMILESN.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more