Brij™-35, 30% Solution
Brij™-35, 30% Solution
Thermo Scientific™

Brij™-35, 30% Solution

This Thermo Scientific formulation of Brij 35 is a stabilized 30% (w/w) solution of Brij-35 detergent for use in variousRead more
Have Questions?
Catalog NumberQuantity
20150950 mL
Catalog number 20150
Price (USD)
201.65
Online Exclusive
233.00
Save 31.35 (13%)
Each
In stock
Add to cart
Quantity:
950 mL
Request bulk or custom format
Price (USD)
201.65
Online Exclusive
233.00
Save 31.35 (13%)
Each
Add to cart
This Thermo Scientific formulation of Brij 35 is a stabilized 30% (w/w) solution of Brij-35 detergent for use in various protein methods. Brij 35 is a nonionic polyoxyethylene surfactant that is most frequently used as a component of cell lysis buffers or a surfactant in various HPLC applications.

Properties of Brij 35 Detergent:

• Molecular Weight: 1225 g
• Detergent Class: Non-ionic
• Aggregation Number: 40
• Micelle Molecular Weight: 49,000 g
• Critical Micelle Concentration (CMC): 0.09 mM (0.011%, w/v)
• Cloud Point: >100°C
• Dialyzable: No

Features of this Thermo Scientific detergent solution:

• A nonionic detergent for use in various protein methods
Accurate—30% detergent solution in ultrapure water
Easy-to-use—solutions are simple to dispense and dilute for use

Specifications of this Thermo Scientific Brij 35 30% solution:

• Visual: Clear, colorless viscous liquid, free of foreign material.
• Concentration: 27.0 to 30.0%


For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionBrij-35, 30% Solution
Quantity950 mL
Reagent TypeDetergent Solution
FormLiquid
Product LineBrij™, Surfact-Amps™
Product TypeDetergent
Unit SizeEach
Contents & Storage
Store at Room Temperature

Save 51% on various cell culture plasticware

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link