Thermo Scientific Chemicals

Copper(I) iodide, 99.995%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 201500250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Copper(I) iodide, 99.995%, (trace metal basis), Thermo Scientific Chemicals

Catalog number: 201500250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 201500250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7681-65-4
IUPAC Name
λ¹-copper(1+) iodide
Molecular Formula
CuI
InChI Key
LSXDOTMGLUJQCM-UHFFFAOYSA-M
SMILES
[Cu+].[I-]
Appearance (Color)
White to grey to light brown
Appearance (Form)
Powder
Total trace metal impurities
=<50 ppm
Titration Complexometric
31.0 to 34.0 % (Cu)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, for ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text