2-Bromo-2-butene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
2-Bromo-2-butene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-2-butene, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 201510250
Price (USD)/ Each
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name(2E)-2-bromobut-2-ene
Molecular FormulaC4H7Br
InChI KeyUILZQFGKPHAAOU-ONEGZZNKSA-N
SMILESC\C=C(/C)Br
Molecular Weight (g/mol)135.00
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4590 to 1.4620 (20°C, 589 nm)
Appearance (Color)Clear yellow to brown
Appearance (Form)Liquid
GC>=97.5 %
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only