Thermo Scientific Chemicals

2-Bromo-2-butene, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 201511000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

2-Bromo-2-butene, 98%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog number: 201511000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 201511000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13294-71-8
IUPAC Name
(2E)-2-bromobut-2-ene
Molecular Formula
C4H7Br
InChI Key
UILZQFGKPHAAOU-ONEGZZNKSA-N
SMILES
C\C=C(/C)Br
Refractive index
1.4590 to 1.4620 (20°C, 589 nm)
Appearance (Color)
Clear yellow to brown
Appearance (Form)
Liquid
GC
>=97.5 %
Appearance
May darken on storage

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text