Thermo Scientific Chemicals

Ammonium persulfate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 201530010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

Ammonium persulfate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 201530010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 201530010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7727-54-0
IUPAC Name
diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular Formula
H8N2O8S2
InChI Key
ROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES
[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Appearance (Color)
White to light yellow
Titration with KMnO4
>=97.5 %
Heavy metals (as Pb)
=<50 ppm
Iron (Fe)
=<10 ppm
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text