Ammonium persulfate, 98%, extra pure, Thermo Scientific Chemicals
Ammonium persulfate, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium persulfate, 98%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 201531000
Price (USD)/ Each
35.65
Online exclusive
42.20 
Save 6.55 (16%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
35.65
Online exclusive
42.20 
Save 6.55 (16%)
Add to cart
Ammonium persulfate, 98%, extra pure, Thermo Scientific Chemicals
Catalog number201531000
Price (USD)/ Each
35.65
Online exclusive
42.20 
Save 6.55 (16%)
-
Add to cart
Chemical Identifiers
IUPAC Namediammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaH8N2O8S2
InChI KeyROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Molecular Weight (g/mol)228.19
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Titration with KMnO4>=97.5 %
Heavy metals (as Pb)=<50 ppm
Appearance (Form)Crystalline powder or crystals
Iron (Fe)=<10 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only