tRNA for LightShift™ Chemiluminescent RNA EMSA Kit
Product Image
Thermo Scientific™

tRNA for LightShift™ Chemiluminescent RNA EMSA Kit

tRNA, formulation: Transfer RNA at 10 mg/mL in 10mM HEPES.Sufficient For: 100 binding reactionsRelated ProductsLightShift™ Chemiluminescent RNA EMSA KitRead more
Have Questions?
Catalog NumberQuantity
20159100 μL
Catalog number 20159
Price (USD)
112.00
Each
Add to cart
Quantity:
100 μL
Request bulk or custom format
Price (USD)
112.00
Each
Add to cart
tRNA, formulation: Transfer RNA at 10 mg/mL in 10mM HEPES.
Sufficient For: 100 binding reactions

Related Products
LightShift™ Chemiluminescent RNA EMSA Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineLightShift™
Product TypetRNA for LightShift RNA EMSA Kit
Quantity100 μL
Target SpecificityNot Target-Specific
Detection MethodChemiluminescence
FormatKit
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -70°C.

Frequently asked questions (FAQs)

What is an electrophoretic mobility shift assay (EMSA)?

EMSAs (also called gel shifts, band shifts, gel retardation assays, or mobility assays) have been used extensively for studying protein-DNA interactions. Because protein-DNA complexes migrate more slowly through a native polyacrylamide or agarose gel than DNA alone, individual protein-DNA complexes can be visualized as discrete bands within the gel using chemiluminescence or radioisotopic detection.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link