Thermo Scientific™

Pierce™ RNA 3' End Biotinylation Kit

Catalog number: 20160
Have Questions?
Thermo Scientific™

Pierce™ RNA 3' End Biotinylation Kit

Catalog number: 20160
Have Questions?
Catalog Number
20160
Unit Size
20 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
2016020 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Pierce RNA 3' End Biotinylation Kit is optimized for labeling the 3-prime terminal end of RNA probes to facilitate their use as probes or targets in EMSA and other methods for studying protein-RNA interactions.

Features of the RNA 3' End Biotinylation Kit:

Non-radioactive—incorporates a biotin label which allows detection sensitivity similar to radioactivity when used with chemiluminescent detection systems
Fast—RNA can be labeled in 0.5-2 hours with minimal downstream processing
Easy to use—RNA ligase and optimized reaction buffer are included
Economical—only a fraction of the cost of purchasing synthetic biotinylated RNA probes
End-labeled—results in minimal disturbance of RNA secondary structure
Flexible—label synthetic or in vitro transcribed RNA probes ranging from 22—450 nucleotides

This complete kit contains an unlabeled RNA strand to use as a positive control, a biotinylated-RNA probe standard, RNase inhibitor, glycogen and ligation-enhancing reagents.

The RNA 3' End Biotinylation Kit uses T4 RNA ligase to conjugate a single nucleotide analog to the 3' terminus of an RNA strand. The unique biotinylated cytidine (bis)phosphate used in this kit enables T4 RNA ligase to attach a single biotin tag on RNA probes of diverse secondary structure and lengths with > 70% efficiency.

Labeling reactions performed with the RNA 3' End Biotinylation Kit typically require 50 pmol of RNA and a 20-fold excess of biotinylated nucleotide in a 2 hour reaction at 16°C. Shorter RNA probes with minimal secondary structure can be ligated in 30 minutes at 37°C (e.g., RNA polymerase RNA template). Large or structurally complex RNA probes (e.g., Let-7 and hTR) require longer incubation times. Optimization of biotinylation efficiency for complex RNA molecules is achieved by altering the RNA to biotinylated cytidine ratio, increasing the incubation time or by adding DMSO to the labeling reaction in order to denature the RNA structure.

Following the RNA biotinylation reaction, labeled RNA is easily precipitated to remove reaction byproducts. The biotin label enables the RNA probe to be immobilized or detected with streptavidin in order to study RNA interactions (i.e., RNA electrophoretic mobility shift assays, RNA pull-down assays) or to perform nonradioactive blotting assays (i.e., Northern blotting and miRNA profiling).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
No. of Reactions
20
Product Line
Pierce™
Product Type
RNA Labeling Kit
Quantity
20 Reactions

Contents & Storage

Sufficient For: 20 labeling reactions (each with 50 pmol RNA)
• T4 RNA Ligase Reaction Buffer (10X), 100 μL
• T4 RNA Ligase (20,000 U/mL), 40 μL
• Non-labeled RNA Control (10 μM), 100 μL
• Biotinylated IRE RNA Control (125 nM), 35 μL
• RNase Inhibitor (40 U/μL), 2 × 10 μL
• Nuclease-free Water, 1.5 mL
• DMSO, 200 μL
• 30% PEG, 300 μL
• Biotinylated Cytidine (Bis)phosphate (1mM), 40 μL
• Glycogen (20 mg/mL), 20 μL

Store all components at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text