Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel, 99.97%, (trace metal basis), spheres, 5 to 11 mm, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantityPackaging
201672500250 gPlastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 201672500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Spheres
Assay>=99.92 % (Ni)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link