Thermo Scientific™

Pierce™ Anti-c-Myc Agarose

Catalog number: 20168
Have Questions?
Thermo Scientific™

Pierce™ Anti-c-Myc Agarose

Catalog number: 20168
Have Questions?
Catalog Number
20168
Unit Size
2 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
201682 mL
-
2016910 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Pierce Anti-c-Myc Agarose is a high-affinity immunoprecipitation resin ideal for purification of recombinant c-Myc-tagged proteins expressed in human in vitro expression systems and bacterial and mammalian cell lysates.

Features of Anti-c-Myc Agarose:

Specific—highly specific anti-c-Myc monoclonal antibody (clone 9E10) enables high yield and high purity for immunoprecipitation
Scalable—available in 2 and 10 mL resin package sizes to allow for larger scale purifications or immunoprecipitations
Versatile—can be used in spin or gravity columns as well as in FPLC cartridges
Convenient—reagents to elute and detect c-Myc tagged fusion proteins are available separately

The anti-c-Myc antibody used to manufacture Pierce Anti-c-Myc Agarose is a highly specific mouse IgG1 monoclonal antibody (clone 9E10) that recognizes the c-Myc epitope tag (EQKLISEEDL) derived from the human c-myc oncogene (p62 c-myc). The support is Superflow 6 resin, a highly crosslinked 6% agarose resin. This anti-c-Myc affinity resin can be packed into gravity purification columns, spin purification columns or cartridges for FPLC instruments to purify c-Myc-fusion proteins expressed in bacterial or mammalian cells.

Applications:
• Purification of c-Myc fusion proteins expressed in the Pierce Human In Vitro Translation kits
• Large scale purification of recombinant c-Myc tagged proteins
• High-throughput enrichment of fusion proteins and interacting partners
• IP and co-IP experiments (as with the Pierce c-Myc-Tag IP/co-IP Kit)

Pierce Anti-c-Myc Agarose consists of a highly specific monoclonal anti-c-Myc antibody that is covalently immobilized on a crosslinked 6% beaded agarose support. The product is supplied as a 25% slurry. Upon incubation with a sample, c-Myc tagged fusion protein is captured on the agarose beads. After simple washing steps, the tagged protein is easily eluted from the resin using 0.1M glycine (pH 2.0-2.8), 3M NaSCN or 50 mM NaOH depending on the downstream application of the purified protein. For elution of highly functional proteins, Pierce c-Myc-peptide can also be used to elute the c-Myc tagged protein. Anti-c-Myc antibody may be used to detect the presence of the tagged protein by Western blot.

In experiments with a c-Myc-tagged fusion protein (26 to 29kDa), the resin provided a binding capacity of 102 to 144nmol protein per mL of settled resin. The elution capacity was at least 18 to 19nmol per mL of settled resin using 0.1M glycine, pH 2.8. Binding and elution capacity will vary depending on the c-Myc-fusion protein and the method of elution.

Related Products
c-Myc Peptide Control for Pierce™ Anti-c-Myc Agarose
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity (Metric)
102 to 144 nmol Protein Binding per mL Resin
Product Line
Pierce™
Target
c-Myc-Tagged Protein
Type
Resin
Stationary Phase
Antibody
Format
Bottle
Column Type
Affinity, Agarose Resin

Contents & Storage

2 mL of settled resin. Provided as 8 mL of 25% slurry.

Store at 4°C. Do not freeze.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text