Thermo Scientific Chemicals

Niobium(V) chloride, 99.8%, Thermo Scientific Chemicals

Catalog number: 201690100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Niobium(V) chloride, 99.8%, Thermo Scientific Chemicals

Catalog number: 201690100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 201690100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10026-12-7
IUPAC Name
niobium(5+) pentachloride
Molecular Formula
Cl5Nb
InChI Key
YHBDIEWMOMLKOO-UHFFFAOYSA-I
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Nb+5]
Silicon (Si)
=<3 ppm
Tin (Sn)
=<1 ppm
Titanium (Ti)
=<3 ppm
Tungsten (W)
=<2 ppm
Zirconium (Zr)
=<1 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text