Niobium(V) chloride, 99.8%, Thermo Scientific Chemicals
Niobium(V) chloride, 99.8%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Niobium(V) chloride, 99.8%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number 201690500
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
Add to cart
Niobium(V) chloride, 99.8%, Thermo Scientific Chemicals
Catalog number201690500
Price (USD)/ Each
103.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
122.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nameniobium(5+) pentachloride
Molecular FormulaCl5Nb
InChI KeyYHBDIEWMOMLKOO-UHFFFAOYSA-I
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Nb+5]
Molecular Weight (g/mol)270.16
View more
SpecificationsSpecification SheetSpecification Sheet
Silicon (Si)=<3 ppm
Tin (Sn)=<1 ppm
Titanium (Ti)=<3 ppm
Tungsten (W)=<2 ppm
Zirconium (Zr)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only