Lead(II) acetate trihydrate, 99+%, for analysis, Thermo Scientific Chemicals
Lead(II) acetate trihydrate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead(II) acetate trihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 201750250
Price (USD)/ Each
26.65
Online exclusive
29.10 
Save 2.45 (8%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
26.65
Online exclusive
29.10 
Save 2.45 (8%)
Add to cart
Lead(II) acetate trihydrate, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number201750250
Price (USD)/ Each
26.65
Online exclusive
29.10 
Save 2.45 (8%)
-
Add to cart
Chemical Identifiers
CAS6080-56-4
IUPAC Nameλ²-lead(2+) diacetate trihydrate
Molecular FormulaC4H12O7Pb
InChI KeyMCEUZMYFCCOOQO-UHFFFAOYSA-L
SMILESO.O.O.[Pb++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Sodium (Na)=<100 ppm
Appearance (Form)Crystalline powder or crystals
Titration Complexometric99.0 to 103.0 %
Chloride (Cl)=<5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only