Ammonium phosphate, dibasic, 99+%, for analysis
Ammonium phosphate, dibasic, 99+%, for analysis
Ammonium phosphate, dibasic, 99+%, for analysis
Ammonium phosphate, dibasic, 99+%, for analysis
Ammonium phosphate, dibasic, 99+%, for analysis
Thermo Scientific Chemicals

Ammonium phosphate, dibasic, 99+%, for analysis

CAS: 7783-28-0 | H9N2O4P | 132.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 201825000
Price (EUR)
54,66
Online Exclusive
64,30
Save 9,64 (15%)
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (EUR)
54,66
Online Exclusive
64,30
Save 9,64 (15%)
Each
Add to cart
Chemical Identifiers
CAS7783-28-0
IUPAC Namephosphoric acid diamine
Molecular FormulaH9N2O4P
InChI KeyMNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILESN.N.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals (as Pb)=<10 ppm
Sodium (Na)=<100 ppm
Appearance (Form)Adhering crystals
Chloride (Cl)=<5 ppm
Sulfate (SO4)=<200 ppm
View more
Ask our AI about this Product

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473850Certificate of AnalysisJun 26, 2025201825000, 201820025, 201822500
A0465529Certificate of AnalysisAug 20, 2024201825000, 201820025, 201822500
A0461980Certificate of AnalysisApr 16, 2024201825000, 201820025, 201822500
A0450539Certificate of AnalysisMay 02, 2023201825000, 201820025, 201822500
A0449151Certificate of AnalysisJan 10, 2023201825000, 201820025, 201822500
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection