Hydrazine sulfate, 99+%, Thermo Scientific Chemicals
Hydrazine sulfate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrazine sulfate, 99+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number 201865000
Price (USD)/ Each
146.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
172.00 
Save 25.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
146.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
172.00 
Save 25.35 (15%)
Add to cart
Hydrazine sulfate, 99+%, Thermo Scientific Chemicals
Catalog number201865000
Price (USD)/ Each
146.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
172.00 
Save 25.35 (15%)
-
Add to cart
Chemical Identifiers
CAS10034-93-2
IUPAC Namehydrazine; sulfuric acid
Molecular FormulaH6N2O4S
InChI KeyZGCHATBSUIJLRL-UHFFFAOYSA-N
SMILESNN.OS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Zinc (Zn)=<5 ppm
Magnesium (Mg)=<5 ppm
Total chloride=<20 ppm
Appearance (Form)Crystalline powder
Calcium (Ca)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only