Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water, Thermo Scientific Chemicals
Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 201950025
Price (USD)/ Each
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS429-41-4, 109-99-9, 7732-18-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to amber to light green to brown
Titration with HClO40.90 to 1.10 M
Water=<8 %
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only