Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water, Thermo Scientific Chemicals
Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number 201955000
Price (USD)/ Each
190.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
224.00 
Save 33.35 (15%)
-
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
190.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
224.00 
Save 33.35 (15%)
Add to cart
Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water, Thermo Scientific Chemicals
Catalog number201955000
Price (USD)/ Each
190.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
224.00 
Save 33.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Nametetrabutylazanium fluoride
Molecular FormulaC16H36FN
InChI KeyFPGGTKZVZWFYPV-UHFFFAOYSA-M
SMILES[F-].CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol)261.47
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO40.90 to 1.10 M
Appearance (Color)Clear yellow to amber to light green to brown
Water=<8 %
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only