Thermo Scientific Chemicals

Hydrazine monohydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 201990051
5 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Hydrazine monohydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 201990051
5 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 201990051
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2644-70-4
IUPAC Name
dihydrogen hydrazine dichloride
Molecular Formula
Cl2H6N2
InChI Key
LIAWOTKNAVAKCX-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NN
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline powder or crystals
Infrared spectrum
Conforms
Titration Argentometric
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text