Hydrazine monohydrochloride, 98+%, Thermo Scientific Chemicals
Hydrazine monohydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrazine monohydrochloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 201992500
Price (USD)/ Each
70.65
Online exclusive
77.90 
Save 7.25 (9%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
70.65
Online exclusive
77.90 
Save 7.25 (9%)
Add to cart
Hydrazine monohydrochloride, 98+%, Thermo Scientific Chemicals
Catalog number201992500
Price (USD)/ Each
70.65
Online exclusive
77.90 
Save 7.25 (9%)
-
Add to cart
Chemical Identifiers
CAS2644-70-4
IUPAC Namedihydrogen hydrazine dichloride
Molecular FormulaCl2H6N2
InChI KeyLIAWOTKNAVAKCX-UHFFFAOYSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration Argentometric>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only