Potassium persulfate, 99+%, for analysis, Thermo Scientific Chemicals
Potassium persulfate, 99+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium persulfate, 99+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
25 g
250 g
500 g
1 kg
Packaging:
Plastic bottle
Catalog number 202010050
Price (USD)/ Each
277.65
Online exclusive
326.00 
Save 48.35 (15%)
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
277.65
Online exclusive
326.00 
Save 48.35 (15%)
Add to cart
Potassium persulfate, 99+%, for analysis, Thermo Scientific Chemicals
Catalog number202010050
Price (USD)/ Each
277.65
Online exclusive
326.00 
Save 48.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaK2O8S2
InChI KeyUSHAGKDGDHPEEY-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Molecular Weight (g/mol)270.31
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Iodometric>=99.0 %
Chloride (Cl)=<120 ppm
Heavy metals (as Pb)=<50 ppm
Iron (Fe)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only