Thermo Scientific Chemicals

Sodium persulfate, +98%, Thermo Scientific Chemicals

Catalog number: 202020050
5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium persulfate, +98%, Thermo Scientific Chemicals

Catalog number: 202020050
5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 202020050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7775-27-1
IUPAC Name
disodium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular Formula
Na2O8S2
InChI Key
CHQMHPLRPQMAMX-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Appearance (Color)
White to almost white
Appearance (Form)
Powder or crystals
Titration Iodometric
98.0 to 102.0 %
Water
=<0.15 %
Heavy metals (as Pb)
=<20 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text